missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mineral Spirits 50000 μg/mL in Dichloromethane, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Distillates (Petroleum), hydrotreated light |
| CAS | 64742-47-8 |
| Analyte or Component Names | Distillates (Petroleum), hydrotreated light |
| Shipping Condition | Room Temperature |
| Concentration | 50000 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Mineral Spirits, Distillates (petroleum), hydrotreated light, Hydrotreated light distillate (petroleum), Dearomatized kerosine, Deodorized kerosene, Low odor paraffinic solvent, UN 1268 |
| Solvent or Matrix | Dichloromethane |
| Quantity | 1 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction