missing translation for 'onlineSavingsMsg'
Learn More

Mineral oil, for spectroscopy, suitable for nujol mull preparations for IR-spect., Thermo Scientific Chemicals

Catalog Number p-169547
14.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
6 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
8042-47-5
C16H10N2Na2O7S2
452.363
MFCD00131611
AEOVEGJBKQQFOP-DDVLFWKVSA-L
acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
9566064
disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
8042-47-5
C16H10N2Na2O7S2
452.363
MFCD00131611
AEOVEGJBKQQFOP-DDVLFWKVSA-L
acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
9566064
disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For IR spectroscopy
TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.8510g/mL
Flash Point 185°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Refractive Index 1.4620 to 1.4730
Quantity 6 mL
Merck Index 14, 7186
Specific Gravity 0.851
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene, chloroform, ether,, carbon dislulfide, petroleum ether, oils, miscible with wax and fats, insoluble in alcohol
Viscosity 14-19 mm2/s (40°C)
Grade Spectroscopy
Physical Form Oily Liquid
Chemical Name or Material Mineral oil
Show More Show Less
compliance-icons
Product Identifier
  • Mineral oil, suitable for nujol mull preparations for IR-spect.
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'