missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mineral Oil, Molecular Biology Grade, Thermo Scientific™
Click to view available options
Quantity:
1 L
Packaging:
Plastic bottle
Specifications
CAS | 8042-47-5 |
Flash Point | >110°C |
Packaging | Plastic bottle |
Molecular Formula | C16H10N2Na2O7S2 |
MDL Number | MFCD00131611 |
Quantity | 1 L |
Synonym | acid orange 10, wool orange 2 g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g |
InChI Key | AEOVEGJBKQQFOP-DDVLFWKVSA-L |
SMILES | C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+] |
IUPAC Name | disodium;(8 Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'