missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mindin/Spondin-2 Antibody (S06-4H7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19780-50ul
This item is not returnable.
View return policy
Description
Mindin/Spondin-2 Monoclonal antibody specifically detects Mindin/Spondin-2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Mindin/Spondin-2 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10417 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| S06-4H7 | |
| Unconjugated | |
| Differentially expressed in cancerous and non-cancerous lung cells 1, DIL-1, DIL1FLJ34460, DKFZp686G21139, FLJ16313, FLJ22401, MINDIN, M-SPONDIN, spondin 2, extracellular matrix protein, spondin-2 | |
| A synthetic peptide of human Mindin/Spondin-2 (Uniprot # Q9BUD6) | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction