missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mind Bomb 2/MIB2 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-68224B
This item is not returnable.
View return policy
Description
Mind Bomb 2/MIB2 Polyclonal antibody specifically detects Mind Bomb 2/MIB2 in Human samples. It is validated for Western Blot
Specifications
| Mind Bomb 2/MIB2 | |
| Polyclonal | |
| Western Blot | |
| E3 ubiquitin-protein ligase MIB2, EC 6.3.2, EC 6.3.2.-, EC 6.3.2.13, FLJ20648, FLJ41406, Mind bomb homolog 2, mindbomb homolog 2 (Drosophila), Novel zinc finger protein, novelzin, Putative NF-kappa-B-activating protein 002N, SKD, Skeletrophin, Zinc finger ZZ type with ankyrin repeat domain protein 1, ZZ type with ankyrin repeat domain 1, ZZANK1 | |
| The immunogen recognized by this antibody maps to a region between residue 925 and 975 of human mindbomb homolog 2 (Skeletrophin) using the numbering given in entry NP_543151.1 (GeneID 142678). | |
| 0.1 mL | |
| Signal Transduction | |
| 142678 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction