missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mind Bomb 1/MIB1 Rabbit anti-Human, Mouse, Rat, Clone: 9Q4N9, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | Mind Bomb 1/MIB1 |
|---|---|
| Clone | 9Q4N9 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18399046
|
Novus Biologicals
NBP3-16175-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18394415
|
Novus Biologicals
NBP3-16175-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mind Bomb 1/MIB1 Monoclonal antibody specifically detects Mind Bomb 1/MIB1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Mind Bomb 1/MIB1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism, Zinc Finger | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 57534 | |
| IgG | |
| Affinity purified |
| 9Q4N9 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DAPK-interacting protein 1, DIP1, DIP-1FLJ90676, DKFZp686I0769, DKFZp761M1710, E3 ubiquitin-protein ligase MIB1, EC 6.3.2, EC 6.3.2.-, KIAA1323, MGC129660, MIB, Mind bomb homolog 1, mindbomb homolog 1 (Drosophila), ubiquitin ligase mind bomb, ubiquitin ligase protein MIB1, Zinc finger ZZ type with ankyrin repeat domain protein 2, ZZANK2MGC129659, ZZZ6 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Mind Bomb 1/MIB1 (Q86YT6). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title