missing translation for 'onlineSavingsMsg'
Learn More

Mind Bomb 1/MIB1 Antibody [DyLight 550], Novus Biologicals™

Product Code. 30731199 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30731199 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731199 Supplier Novus Biologicals Supplier No. AF7305L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Mind Bomb 1/MIB1 Polyclonal antibody specifically detects Mind Bomb 1/MIB1 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mind Bomb 1/MIB1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias DAPK-interacting protein 1, DIP1, DIP-1FLJ90676, DKFZp686I0769, DKFZp761M1710, E3 ubiquitin-protein ligase MIB1, EC 6.3.2, EC 6.3.2.-, KIAA1323, MGC129660, MIB, Mind bomb homolog 1, mindbomb homolog 1 (Drosophila), ubiquitin ligase mind bomb, ubiquitin ligase protein MIB1, Zinc finger ZZ type with ankyrin repeat domain protein 2, ZZANK2MGC129659, ZZZ6
Host Species Sheep
Immunogen E. coli-derived recombinant human Mind Bomb 1/MIB1, Thr365-Gln461, Accession # Q86YT6
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 57534
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.