missing translation for 'onlineSavingsMsg'
Learn More

MINA Antibody (768826) [DyLight 550], Novus Biologicals™

Product Code. 30656399 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30656399 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30656399 Supplier Novus Biologicals Supplier No. FAB7266L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

MINA Monoclonal antibody specifically detects MINA in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MINA
Applications Western Blot
Classification Monoclonal
Clone 768826
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias DKFZp762O1912, FLJ14393, MDIGDKFZp762O1912, MINA53mdig, mineral dust induced gene protein, Mineral dust-induced gene protein, MYC induced nuclear antigen, MYC-induced nuclear antigen, myc-induced nuclear antigen, 53 kDa, NO52, Nucleolar protein 52
Host Species Rat
Immunogen E. coli-derived recombinant mouse MINA, Met1-Gly192, Accession # Q8CD15
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 84864
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.