missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MINA Antibody (753002) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7476JF585
This item is not returnable.
View return policy
Description
MINA Monoclonal antibody specifically detects MINA in Human samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| MINA | |
| Monoclonal | |
| Janelia Fluor 585 | |
| DKFZp762O1912, FLJ14393, MDIGDKFZp762O1912, MINA53mdig, mineral dust induced gene protein, Mineral dust-induced gene protein, MYC induced nuclear antigen, MYC-induced nuclear antigen, myc-induced nuclear antigen, 53 kDa, NO52, Nucleolar protein 52 | |
| E. coli-derived recombinant human MINA, Met1-Gly192, Accession # Q8IUF8 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 753002 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 84864 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction