missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MINA Antibody (1E8), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | MINA |
|---|---|
| Clone | 1E8 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30027554
|
Novus Biologicals
NBP3-26342-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30036057
|
Novus Biologicals
NBP3-26342-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MINA Monoclonal antibody specifically detects MINA in Human samples. It is validated for ELISASpecifications
| MINA | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp762O1912, FLJ14393, MDIGDKFZp762O1912, MINA53mdig, mineral dust induced gene protein, Mineral dust-induced gene protein, MYC induced nuclear antigen, MYC-induced nuclear antigen, myc-induced nuclear antigen, 53 kDa, NO52, Nucleolar protein 52 | |
| A synthesized peptide derived from Human MINA [UniProt Q8IUF8] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 1E8 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| DNA replication Transcription Translation and Splicing | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 84864 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title