missing translation for 'onlineSavingsMsg'
Learn More

Miltefosine, MedChemExpress

Product Code. 16411379
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
This item is not returnable. View return policy

Product Code. 16411379

Brand: Medchem Express HY13685

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Miltefosine is a broad spectrum antimicrobial, anti-leishmanial, phospholipid agent acting by inhibiting the PI3K/Akt activity. Miltefosine is an inhibitor of CTP-phosphocholine cytidyltransferase (CCT).

Miltefosine is a broad spectrum antimicrobial, anti-leishmanial, phospholipid agent acting by inhibiting the PI3K/Akt activity. Miltefosine is an inhibitor of CTP-phosphocholine cytidyltransferase (CCT).

Specifications

Chemical Name or Material Miltefosine
Physical Form Solid
CAS 58066-85-6
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C21H46NO4P
For Use With (Application) COVID-19-anti-virus
Synonym HePC Hexadecyl phosphocholine
Solubility Information H2O : 50 mg/mL (122.68 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (12.27 mM; Need ultrasonic)
SMILES O=P(OCC[N+](C)(C)C)(OCCCCCCCCCCCCCCCC)[O-]
Molecular Weight (g/mol) 407.57
Formula Weight 407.57
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.