missing translation for 'onlineSavingsMsg'
Learn More

Milrinone, Tocris Bioscience™
GREENER_CHOICE

Product Code. 30001374
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001374

Brand: Tocris Bioscience 1504/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

PDE3 inhibitor

Potent cAMP phosphodiesterase inhibitor (IC50 = 56 nM for inhibition of PDE III). Has inotropic and vasodilator effects following oral or intravenous administration in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78415-72-2
Molecular Formula C12H9N3O
Molecular Weight (g/mol) 211.224
InChI Key PZRHRDRVRGEVNW-UHFFFAOYSA-N
Synonym milrinone, primacor, corotrope, corotrop, milrinona, milrinonum, milrinonum latin, milrinona spanish, milrila, unii-ju9yax04c7
PubChem CID 4197
ChEBI CHEBI:50693
IUPAC Name 6-methyl-2-oxo-5-pyridin-4-yl-1H-pyridine-3-carbonitrile
SMILES CC1=C(C=C(C(=O)N1)C#N)C2=CC=NC=C2

Specifications

Quantity 10 mg
Formula Weight 211.22
Percent Purity >98%
Chemical Name or Material Milrinone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.