missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Milrinone, 97%

Product Code. 10555154 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
Packaging:
Glass bottle
Unit Size:
250mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10555154 50 mg Glass bottle 50mg
10215074 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10555154 Supplier Thermo Scientific Chemicals Supplier No. 397870500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78415-72-2
Molecular Formula C12H9N3O
Molecular Weight (g/mol) 211.22
MDL Number MFCD00133539
InChI Key PZRHRDRVRGEVNW-UHFFFAOYSA-N
Synonym milrinone, primacor, corotrope, corotrop, milrinona, milrinonum, milrinonum latin, milrinona spanish, milrila, unii-ju9yax04c7
PubChem CID 4197
ChEBI CHEBI:50693
IUPAC Name 6-methyl-2-oxo-5-pyridin-4-yl-1H-pyridine-3-carbonitrile
SMILES CC1=C(C=C(C(=O)N1)C#N)C2=CC=NC=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point >280°C
Color Pink to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 50 mg
Merck Index 15, 6278
Formula Weight 211.22
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Milrinone
Show More Show Less
compliance-icons
Product Identifier
  • Milrinone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
Hazard Statement
  • H330-Fatal if inhaled.
  • H311-Toxic in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.