missing translation for 'onlineSavingsMsg'
Learn More

Merck Stopper for Vacuum Filtration

Product Code. 15879512
Change view
Click to view available options
Quantity:
5 Pk.
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15879512 5 Pk. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15879512 Supplier Merck Supplier No. XX1014708
  • ONLINE EXCLUSIVE PRICE
    £258.00
    £157.39 / Pack of 5
    Save £100.61
    38% Off
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stopper No. 8 is designed to replace parts contained in glass vacuum filtration kits or provide for an alternative configuration.

Millipore Glass Vacuum Filtration Accessory Product:

  • Stopper, No. 8, perforated, parylene coated, 5 Pk.
  • These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.
  • This item is an accessory part for the Millipore 47 mm Classic Glass Filter Holder Kits.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material Silicone
For Use With (Application) Vacuum filtration
Quantity 5 Pk.
Diameter (Metric)Top 41.2 mm
For Use With (Equipment) Millipore Classic Glass Filter Holder Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.