missing translation for 'onlineSavingsMsg'
Learn More

Merck Spring Clamp for Vacuum Filtration

Product Code. 15874156 Shop All Merck Products
Click to view available options
Color:
Clear
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15874156

Brand: Merck XX1014703

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Designed to replace parts contained in glass vacuum filtration kits or provide for an alternative configuration

Millipore Glass Vacuum Filtration Accessory Product

Spring Clamp, 47 mm, Aluminum

These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.

This item is an accessory part for the Millipore 47 mm Classic Glass Filter Holder Kits, Millipore 47 mm All-Glass Filter Holder Kits and Millipore MilliSolve Kit.

TRUSTED_SUSTAINABILITY

Specifications

Type Laboratory Filter Holders or Cyclones
Material Anodized Aluminum
For Use With (Equipment) Millipore Classic Glass Filter Holder Kits, All-Glass Filter Holder Kits and Millipore MilliSolve Ki
For Use With (Application) Vacuum filtration
Diameter (Metric) 47 mm
Color Clear
Diameter (Metric) Inside 47 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.