missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Cell growth, structure, and function more closely mimic what occurs in vivo
- In addition, Millicell inserts make it possible to study both sides of the cell monolayer
- The inserts are easily prepared for SEM and TEM visualizing techniques, and they are compatible with cellular and/or fluorescent stains
- For co-culturing and permeability assays
- Unique design allows easier basolateral access than other hanging inserts with less risk of contamination
- Available in 5 pore sizes and 3 diameters, including a 1 μm pore size that is optically transparent for better visualization by microscopy
- PET Membrane (polyethylene terephthalate) for growth of attachment-dependent cells without matrix
- Individually blister packaged
Specifications
Specifications
| Description | PET (Polyethylene Terephthalate) Filter Code |
| Material | Polystyrene |
| Pore Size | 3 μm |
| Type | Cell Culture Insert |
| Item Description | 50°C Max. Operating Temperature |
| For Use With (Application) | Cell Attachment, Cell Culture, Cell Differentiation, Immunocytochemistry |
| Diameter (Metric) Filter | 24 mm |
| Quantity | 48/Pk |
| For Use With | 6-well Plates |
| Color | Clear |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?