Learn More
MilliporeSigma Millicell™ ERS 3.0 Digital Voltohmmeter
Product Code. 18844620
Change view
Click to view available options
Voltage:
0/1 V
Unit Size:
Each
Product Code. Voltage Unit Size
18844620 0/1 V Each
1 options
Product Code. 18844620 Supplier MilliporeSigma Supplier No. MERS03000
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Millicell Electrical Resistance System reliably measures transmembrane potential and resistance of endothelial/epithelial cells in culture.
  • Reliably measures membrane potential and resistance of epithelial and endothelial cells in culture
  • Qualitatively measures cell monolayer health and cell confluence
  • An Electroless Nickle Immersion Gold (ENIG) plate on each external facing electrode tip measures voltage
  • Small electrode size enables easy measurement of transepithelial voltage and the resistance of cells grown on microporous membranes
  • Comes with the Voltohmmeter, standard adjustable electrode, a Wi-Fi USB dongle, a power cord, a verification device and free 6-month trial subscription to Millicell™ Cloud account.
  • Each instrument is battery compatible but can run off only AC wall power in the absence of a battery.
TRUSTED_SUSTAINABILITY
Specifications
Type Digital Voltohmmeter
Voltage 0/1 V