missing translation for 'onlineSavingsMsg'
Learn More

Merck Millex™-HV Sterile Syringe Filter Unit, PVDF, 0.45 μm
BIOPHARMA

Product Code. 10487342 Shop All Merck Products
Change view
Click to view available options
Type:
Syringe Filter
Syringe Filter (Medical)
Diameter (Metric):
4 mm
13 mm
33 mm
Quantity:
100/Pk.
250/Pk.
50/Pk.
Unit Size:
Pack of 100
Pack of 250
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Diameter (Metric) Quantity unitSize
10487342 Syringe Filter 33 mm 50/Pk. Pack of 50
10785534 Syringe Filter (Medical) 33 mm 50/Pk. Pack of 50
10030172 Syringe Filter 33 mm 250/Pk. Pack of 250
10156630 Syringe Filter 4 mm 100/Pk. Pack of 100
10146520 Syringe Filter 13 mm 100/Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10487342 Supplier Merck Supplier No. SLHV033RS

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Ready-to-use, syringe-driven units designed for sterilizing and clarifying sterile solutions are available in 0.1, 0.22, 0.45 and 5.0μm pore sizes

Sterile Millex filter units are available in a larger 33 mm housing with either MCE, PVDF or PES membranes.Faster flow rate:The larger filter surface area increases flow rate and throughput. It also makes it easier to filter solutions because it reduces the pressure required to empty the syringe.Higher operating pressure:Millex filters have a maximum housing pressure of 10.5 bar (150 psig), which means you can filter solutions faster than before.Color-coded:The color-coded band on the Millex housing clearly indicates which membrane is inside.Manufactured for reliable performance:Millex filters are manufactured in a controlled environment using an automated process. Human hands never touch the filter during assembly. A Certificate of Quality included in each box describes Millipore's quality standards in detail.Choice of membranes:Millex filters are available with three membranes: Millipore Express™ (PES) membrane, mixed cellulose esters (MCE), and Durapore (PVDF). - Millex-GP filters feature Millipore Express (PES) membrane, which provides a unique combination of speed and six times less protein binding than competitive PES membranes. Millex-GP filters are recommended for sterile filtering protein solutions, tissue culture media, buffers and additives. - Millex filters with Durapore™ (PVDF) membrane are the lowest protein-binding syringe filters available. Use them to filter protein solutions. The 0.2 μm filters are sterilizing grade. Larger pore sizes are for clarification and prefiltration. - Millex-MF filters feature mixed cellulose esters (MCE) membrane, which provides reliable, general purpose filtration of water or other aqueous solutions. The 0.2 μm filters are sterilizing grade. Larger pore sizes are for clarification and prefiltration.Applications:Sterilization of Aqueous Solutions, Organic Solvents and Gases, Filtration of High Value Protein or Small Molecule Components.
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance Meets USP Class VI Biological tests for plastics. All 33 mm filters are CE-marked and suitable for both medical and laboratory applications outside the U.S.
Color Yellow
Disposable Single-use
Filter Type Membrane Filter
Material (Membrane) PVDF (Hydrophilic)
Pore Size 0.45 μm
Sterility Sterile
For Use With (Application) Ultra-low protein binding; Sterilization of tissue culture media, protein solutions or aqueous solutions
Hold up Volume (After Air Purge) <100 μL
Housing Material PVDF
Inlet Style Female Luer Lock
Outlet Style Male Luer Slip
Pressure Rating 145 psi
Quantity 50/Pk.
Temperature (English) Operating 113°F
Temperature (Metric) Operating 45°C
Temperature Range (English) 113°F
Temperature Range (Metric) 45°C
Tip Material Modified Acrylic
Type Syringe Filter
Volume (Metric) 100 mL
Diameter (Metric) 33 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.