missing translation for 'onlineSavingsMsg'
Learn More

Melford Millers LB Broth 1G Capsules

Product Code. 30018409
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
This item is not returnable. View return policy

Product Code. 30018409

Brand: Melford L24041500.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High salt formula.

High salt formula. Molecular Biology Grade. For maintenance and propagation of E. Coli.

RPI Media is now available in convenient pre-measured and pre-mixed capsules. Prebuffered media eliminates messy weighing and pH adjustments.

Simply dissolve 25 capsules per 1 Liter of distilled water. Water must be 37°C or warmer to completely dissolve gelatin capsule. Agitate. Sterilize by autoclaving for 15 minutes before use. Buffer is set for 7.0± 0.2 at 25°C using a 10mM solution of Tris/Tris HCl. pH will vary depending on temperature.

Ingredients:

Casein digest of peptone: 10 g/L

Sodium Chloride: 10 g/L

Yeast Extract: 5 g/L

Tris/Tris HCl: 1.5 g/L

Specifications

Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.