missing translation for 'onlineSavingsMsg'
Learn More

Melford Millers LB Agar Granulated

Product Code. 30018392
Click to view available options
Quantity:
100 g
500 g
1 kg
2 kg
Unit Size:
100g
1kg
2kg
500g
This item is not returnable. View return policy

Product Code. 30018392

Brand: Melford L24022100.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For propagation and maintenance of E°.

For propagation and maintenance of E°. Coli.

Dissolve 37 grams in 1 Liter distilled water and adjust the pH to 7.2. Sterilize by autoclaving for 15 minutes.

Ingredients:

Tryptone: 10g/l

Sodium Chloride: 10g/l

Yeast Extract: 5g/l

Agar: 12g/l

Specifications

Quantity 100 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.