missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Luria-Bertani Agar, pre buffered capsules

Product Code. 12871670
Change view
Click to view available options
Quantity:
500 g
25 x 20 g
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12871670 500 g Each
12861670 25 x 20 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12871670 Supplier Fisher BioReagents™ Supplier No. 12871670
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Used for culturing E. coli.

  • Pre-buffered

Specifications

Content And Storage Room Temperature
Format Capsules
Product Type Agar
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.