Learn More
Melford Miller's LB Agar, 20G Capsules
Description
RPI media is now available in convenient premeasured and pre-mixed capsules.
RPI media is now available in convenient premeasured and pre-mixed capsules. Prebuffered media eliminates pH adjustments. Capsules contain all necessary ingredients for maintenance and propagation of E. coli.
Simply add 2 x 500 mL capsules to one liter of water, sterilize by autoclaving for 15 minutes at 121°C. Supplied as 40 gm capsule.
Final Solution Contains:
Tryptone: 10 g/l
Yeast Extract: 5 g/l
Sodium Choride: 10 g/l
Agar: 15 g/l
Tris/Tris HCL: 1.5 g/l
Specifications
Specifications
| Quantity | 1 kg |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.