missing translation for 'onlineSavingsMsg'
Learn More

Melford Miller's LB Agar, 1G Capsules

Product Code. 30018390
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
This item is not returnable. View return policy

Product Code. 30018390

Brand: Melford L240211000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Super convenient.

Super convenient. Eliminate messy weighing. Consistant and accurate measurement. Convenient premeasured and pre-mixed capsules. Prebuffered media eliminates pH adjustments.

Simply add 40 capsules to one liter of water, sterilize by autoclaving for 15 minutes at 121°C. Capsules contain all necessary ingredients for maintenance and propagation of E. Coli.

Ingredients:

Casein Digest Peptone: 10g/L

Yeast Extract: 5g/L

Sodium Chloride: 10g/L

Agar:15g/L

TRIS/TRIS HCl: 1.5g/L

Specifications

Quantity 1 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.