missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Miller's LB Agar, High Salt, Granulated

Product Code. 12811660
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12811660 2 kg 2kg
12821660 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12811660 Supplier Fisher BioReagents™ Supplier No. 12811660

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For propagation and maintenance of E. coli.

  • Specially granulated to reduce dust and increase solubility range
  • Soluble in water

Specifications

Content And Storage Room Temperature
Form Granulated Powder
Product Type LB Agar
For Use With (Application) For propagation and maintenance of E. Coli.
Quantity 2 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.