missing translation for 'onlineSavingsMsg'
Learn More

Milbemycin Oxime (Mixture of Milbemycin Oxime A3 and Milbemycin Oxime A4, 80:20), TRC

Product Code. 30135352
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135352 10 mg 10mg
30135329 50 mg 50mg
18051120 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135352 Supplier Toronto Research Chemicals Supplier No. TRCM34428010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129496-10-2
Molecular Formula C31H43NO7 and C32H45NO7
Molecular Weight (g/mol) 1097.38
Synonym Milbemycin Oxime, Milbemycin oximes, Safeheart
IUPAC Name (2R,2a'E,2a1'S,4'E,6R,6'R,8'E,11'R,15'S,17a'R,20'Z,20a'R)-2a1'-hydroxy-20'-(hydroxyimino)-5,6,6',8',19'-pentamethyl-2a1',3,4,5,6,6',7',10',11',14',15',17a',20',20a'-tetradecahydro-2'H,17'H-spiro[pyran-2,13'-[11,15]methano[1,5]dioxacyclooctadecino[9,8,7-cd]benzofuran]-17'-one--(2R,2a'E,2a1'S,4'E,6R,6'R,8'E,11'R,15'S,17a'R,20'Z,20a'R)-6-ethyl-2a1'-hydroxy-20'-(hydroxyimino)-5,6',8',19'-tetramethyl-2a1',3,4,5,6,6',7',10',11',14',15',17a',20',20a'-tetradecahydro-2'H,17'H-spiro[pyran-2,13'-[11,15]methano[1,5]dioxacyclooctadecino[9,8,7-cd]benzofuran]-17'-one (4/1)

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C32H45NO7.C31H43NO7/c1-6-27-21(4)12-13-31(40-27)17-25-16-24(39-31)11-10-20(3)14-19(2)8-7-9-23-18-37-29-28(33-36)22(5)15-26(30(34)38-25)32(23,29)35;1-18-7-6-8-23-17-36-28-27(32-35)21(4)14-26(31(23,28)34)29(33)37-25-15-24(10-9-19(2)13-18)39-30(16-25)12-11-20(3)22(5)38-30/h7-10,15,19,21,24-27,29,35-36H,6,11-14,16-18H2,1-5H3;6-9,14,18,20,22,24-26,28,34-35H,10-13,15-17H2,1-5H3/b8-7+,20-10+,23-9+,33-28-;7-6+,19-9+,23-8+,32-27-/t19?,21?,24-,25+,26+,27-,29-,31-,32-;18?,20?,22-,24-,25+,26+,28-,30+,31-/m11/s1
Formula Weight 1096.6236
Chemical Name or Material Milbemycin Oxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.