missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Milbemycin oxime, MedChemExpress
Description
Milbemycin oxime is a macrocyclic lactone and has broad-spectrum anti-parasitic activity. Milbemycin oxime is composed of milbemycins A4 and A3. Milbemycin oxime binds glutamate-gated chloride channels. Milbemycin oxime is against intestinal nematodes, pulmonary and cardiac helminths.
Specifications
Specifications
| Chemical Name or Material | Milbemycin oxime |
| Physical Form | Solid |
| CAS | 129496-10-2 |
| Color | White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C63H88N2O14 |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | DMSO : ≥ 100 mg/mL (91.13 mM) ∣H2O : 1 mg/mL (0.91 mM; ultrasonic and warming and heat to 80°C) |
| SMILES | CC1=C[C@@H]2[C@@]3(O)[C@H](OC/C3=C\C=C\[C@H](C)C/C(C)=C/C[C@@H](O[C@@]4(CC[C@H](C)[C@@H](C)O4)C5)C[C@@H]5OC2=O)/C1=N\O.CC6=C[C@@H]7[C@@]8(O)[C@H](OC/C8=C\C=C\[C@H](C)C/C(C)=C/C[C@@H](O[C@@]9(CC[C@H](C)[C@@H](CC)O9)C%10)C[C@@H]%10OC7=O)/C6=N\O |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?