missing translation for 'onlineSavingsMsg'
Learn More

Milademetan, MedChemExpress

Product Code. 16493308
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493308 10 mg 10mg
16416037 5 mg 5mg
16410749 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16493308 Supplier Medchem Express Supplier No. HY101266

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Milademetan (DS-3032) is a specific and orally active MDM2 inhibitor for the research of acute myeloid leukemia (AML) or solid tumors. Milademetan (DS-3032) induces G1 cell cycle arrest, senescence and apoptosis.

Milademetan (DS-3032) is a specific and orally active MDM2 inhibitor for the research of acute myeloid leukemia (AML) or solid tumors. Milademetan (DS-3032) induces G1 cell cycle arrest, senescence and apoptosis.

Specifications

Chemical Name or Material Milademetan
CAS 1398568-47-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H34Cl2FN5O4
Quantity 10 mg
Synonym DS-3032
Solubility Information DMSO : 16.67 mg/mL (26.95 mM; Need ultrasonic)
SMILES O=C(NC1=CC(Cl)=CC=C21)[C@]32C4(CCC(C)(C)CC4)N[C@@H](C(N[C@@H]5CC[C@@H](C(N)=O)OC5)=O)[C@@H]3C6=CC=NC(Cl)=C6F
Molecular Weight (g/mol) 618.53
Formula Weight 618.53
Percent Purity 92.38%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.