missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MIIP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100572
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total IIP45.
MIIP inhibits glioma cells invasion and down-regulates adhesion-and motility-associated genes such as NFKB2 and ICAM1. MIIP exhibits opposing effects to IGFBP2 on cell invasion.
Specifications
| MIIP | |
| Polyclonal | |
| Unconjugated | |
| MIIP | |
| AA553001; AI853621; D4Wsu114e; IGFBP2-binding protein; IGFBP-2-Binding Protein, IIp45; Iip45; invasion inhibitory protein 45; invasion-inhibitory protein 45; migration and invasion inhibitory protein; migration and invasion-inhibitory protein; MIIP | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 28010, 60672 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A2A7Y5, Q5JXC2 | |
| MIIP | |
| A synthesized peptide derived from human MIIP(Accession Q5JXC2), corresponding to amino acid residues E104-K154. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction