missing translation for 'onlineSavingsMsg'
Learn More

Miglitol, TRC

Product Code. 30138651
Change view
Click to view available options
Quantity:
10 mg
25 mg
1 g
Unit Size:
10mg
1g
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138651 1 g 1g
30132868 25 mg 25mg
17958674 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138651 Supplier Toronto Research Chemicals Supplier No. TRCM3442001G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72432-03-2
Molecular Formula C8 H17 N O5
Molecular Weight (g/mol) 207.22
Synonym 3,4,5-Piperidinetriol, 1-(2-hydroxyethyl)-2-(hydroxymethyl)-, (2R,3R,4R,5S)- (9CI, ACI), (2R,3R,4R,5S)-1-(2-Hydroxyethyl)-2-(hydroxymethyl)-3,4,5-piperidinetriol (ACI), 3,4,5-Piperidinetriol, 1-(2-hydroxyethyl)-2-(hydroxymethyl)-, [2R-(2α,3β,4α,5β)]- (ZCI), (2R,3R,4R,5S)-1-(2-Hydroxyethyl)-2-(hydroxymethyl)piperidine-3,4,5-triol, BAY 1099, BAY-m 1099, Diastabol, Glyset, Miglitol, Miglitose, N-(2-Hydroethyl)deoxynojirimycin, N-(2-Hydroxyethyl)-1-deoxynojirimycin, N-(2-Hydroxyethyl)moranoline, Plumarol, Seibule
IUPAC Name (2R,3R,4R,5S)-1-(2-hydroxyethyl)-2-(hydroxymethyl)piperidine-3,4,5-triol
SMILES OCCN1C[C@H](O)[C@@H](O)[C@H](O)[C@H]1CO

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H17NO5/c10-2-1-9-3-6(12)8(14)7(13)5(9)4-11/h5-8,10-14H,1-4H2/t5-,6+,7-,8-/m1/s1
Formula Weight 207.1107
Chemical Name or Material Miglitol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.