missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer Mighty Small precast gels
Click to view available options
Gel Percentage:
10%
12%
4 to 12%
4 to 20%
8%
Wells:
12-well
15-well
Unit Size:
Pack of 10
Description
- Specification:
- Plate Size: 10 x 8 cm
- Gel Size: 8.5 x 7 cm, 1 mm thickness
- Wells: 15 well (12 wells available)
- System/Buffer: Bis-Tris System, MOPS Buffer
- Loading Volume: For 15 wells: max 30 μl (recommended: ≤ 15 μl)
- Package: 10 pcs/box
- Weight:0.66 lb.
- Storage temp: 2 to 8°
- Shelf life: 12 months
- Advantages:
- upgraded Bis-tris system with moderate PH to ensure less modification on the protein to get a sharp and clear band
- Save trouble and time from making gels
Specifications
Specifications
| Buffer | MOPS |
| Color | Clear |
| Concentration | 8% |
| For Use With (Equipment) | Mini Gel Tank |
| Gel Percentage | 8% |
| Gel Size | Mini |
| Gel Thickness | 1.0 mm |
| Gel Type | Bis-Tris |
| pH Range | 7 |
| Product Type | Precast gels |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion