missing translation for 'onlineSavingsMsg'
Learn More

Migfilin Antibody (FBLIM1/4600), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18390837 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390837 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18390837 Supplier Novus Biologicals Supplier No. NBP314149AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Migfilin Monoclonal antibody specifically detects Migfilin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Migfilin
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone FBLIM1/4600
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CAL, DKFZp434G171, FBLP-1FBLP1CSX-associated LIM, filamin binding LIM protein 1, filamin-binding LIM protein 1, filamin-binding LIM protein-1, MIG2-interacting protein, Migfilin, mitogen-inducible 2 interacting protein, Mitogen-inducible 2-interacting protein
Host Species Mouse
Immunogen Recombinant full-length human Migfilin protein (Uniprot: Q8WUP2)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54751
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.