missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIG6/ERRFI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11630-1-AP-150UL
This item is not returnable.
View return policy
Description
ERRFI1 is a cytoplasmic protein whose expression is upregulated with cell growth (Wick et al., 1995 ). It shares significant homology with the protein product of rat gene-33, which is induced during cell stress and mediates cell signaling (Makkinje et al., 2000 ; Fiorentino et al., 2000 ).Specifications
| MIG6/ERRFI1 | |
| Polyclonal | |
| Unconjugated | |
| ERRFI1 | |
| ERRFI1, GENE 33, MIG 6, MIG6, RALT | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 313729, 54206, 74155 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P05432, Q99JZ7, Q9UJM3 | |
| Errfi1 | |
| MIG6; ERRFI1 Fusion Protein Ag2202 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction