missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIG2/Kindlin-2 Mouse anti-Human, Mouse, Rat, Bovine, Hamster, Clone: 3A3, Novus Biologicals™
Mouse Monoclonal Antibody
£197.00 - £396.00
Specifications
| Antigen | MIG2/Kindlin-2 |
|---|---|
| Clone | 3A3 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18308352
|
Bio-Techne
NBP3-13846-20UG |
20 μg |
£197.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18362892
|
Novus Biologicals
NBP3-13846-100UG |
100 μg |
£396.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MIG2/Kindlin-2 Monoclonal antibody specifically detects MIG2/Kindlin-2 in Human, Mouse, Rat, Bovine, Hamster samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| MIG2/Kindlin-2 | |
| 0.2 mg/ml | |
| Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat, Bovine, Hamster | |
| Q96AC1 | |
| 10979 | |
| IgG1 κ | |
| Protein A or G purified |
| 3A3 | |
| Western Blot 1-2 ug/ml, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| fermitin family homolog 2, fermitin family homolog 2 (Drosophila), fermitin family member 2, KIND2DKFZp686G11125, kindlin 2, Kindlin-2, mig-2, MIG2FLJ34213, mitogen inducible gene 2 protein, Mitogen-inducible gene 2 protein, PH domain-containing family C member 1, pleckstrin homology domain containing, family C (with FERM domain) member 1, pleckstrin homology domain containing, family C member 1, Pleckstrin homology domain-containing family C member 1, PLEKHC1FLJ44462, UNC112, UNC112B | |
| Recombinant protein containing MIG2/Kindlin-2 (residues 218-486) (Uniprot: Q96AC1) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title