missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIG2/Kindlin-2 Antibody (3A3), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-14275PE
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MIG2/Kindlin-2 Monoclonal antibody specifically detects MIG2/Kindlin-2 in Human, Mouse, Rat, Bovine, Hamster samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Especificaciones
| MIG2/Kindlin-2 | |
| Monoclonal | |
| PE | |
| fermitin family homolog 2, fermitin family homolog 2 (Drosophila), fermitin family member 2, KIND2DKFZp686G11125, kindlin 2, Kindlin-2, mig-2, MIG2FLJ34213, mitogen inducible gene 2 protein, Mitogen-inducible gene 2 protein, PH domain-containing family C member 1, pleckstrin homology domain containing, family C (with FERM domain) member 1, pleckstrin homology domain containing, family C member 1, Pleckstrin homology domain-containing family C member 1, PLEKHC1FLJ44462, UNC112, UNC112B | |
| Recombinant protein containing MIG2/Kindlin-2 (residues 218-486) (Uniprot: Q96AC1) | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Bovine, Hamster | |
| Purified |
| Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 3A3 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 10979 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido