missing translation for 'onlineSavingsMsg'
Learn More

MIG2/Kindlin-2 Antibody (3A3), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18394534 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394534 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394534 Supplier Novus Biologicals Supplier No. NBP314275AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MIG2/Kindlin-2 Monoclonal antibody specifically detects MIG2/Kindlin-2 in Human, Mouse, Rat, Bovine, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIG2/Kindlin-2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3A3
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias fermitin family homolog 2, fermitin family homolog 2 (Drosophila), fermitin family member 2, KIND2DKFZp686G11125, kindlin 2, Kindlin-2, mig-2, MIG2FLJ34213, mitogen inducible gene 2 protein, Mitogen-inducible gene 2 protein, PH domain-containing family C member 1, pleckstrin homology domain containing, family C (with FERM domain) member 1, pleckstrin homology domain containing, family C member 1, Pleckstrin homology domain-containing family C member 1, PLEKHC1FLJ44462, UNC112, UNC112B
Host Species Mouse
Immunogen Recombinant protein containing MIG2/Kindlin-2 (residues 218-486) (Uniprot: Q96AC1)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10979
Target Species Human, Mouse, Rat, Bovine, Hamster
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.