missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIG Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22355-1-AP-20UL
This item is not returnable.
View return policy
Description
MIG, also known as CXCL9, is a T-cell chemoattractant inducible by gamma interferon that is a member of the CXC chemokine family of cytokines.This gene is mapped to 4q21. It is noted that, although the best-described activities of the chemokines are as chemotactic factors, chemokines also have an effect on T-cell activation, angiogenesis, and HIV infection. While most CXC chemokines are chemotactic for neutrophils, MIG and INP10 are unusual and similar in being CXC chemokines that are chemotactic for lymphocytes and inactive in neutrophils.Specifications
| MIG | |
| Polyclonal | |
| Unconjugated | |
| CXCL9 | |
| C X C motif chemokine 9, CMK, crg 10, CXCL9, Humig, MIG, MIG; CXCL9, SCYB9, Small inducible cytokine B9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4283 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q07325 | |
| CXCL9 | |
| MIG Fusion Protein Ag17932 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction