missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mig-6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550708
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total Mig-6 protein.
ERRFI1 is a cytoplasmic protein whose expression is upregulated with cell growth (Wick et al., 1995). It shares significant homology with the protein product of rat gene-33, which is induced during cell stress and mediates cell signaling (Makkinje et al., 2000).
Specifications
| Mig-6 | |
| Polyclonal | |
| Unconjugated | |
| Errfi1 | |
| 1300002F13Rik; 33; AI788755; ERBB receptor feedback inhibitor 1; Errfi1; gene 33; Gene 33 polypeptide; GENE-33; MIG6; MIG-6; Mitogen-inducible gene 6 protein; mitogen-inducible gene 6 protein homolog; RALT; Receptor-associated late transducer; RGD1307599; Xxx | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54206 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2.3 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9UJM3 | |
| Errfi1 | |
| Synthetic peptide corresponding to a region derived from internal residues of human ERBB receptor feedback inhibitor 1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction