missing translation for 'onlineSavingsMsg'
Learn More

Mifepristone, MedChemExpress

Product Code. 16491686
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491686 500 mg 500mg
16476276 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16491686 Supplier Medchem Express Supplier No. HY13683
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Mifepristone (RU486) is a progesterone receptor (PR) and glucocorticoid receptor (GR) antagonist with IC50s of 0.2 nM and 2.6 nM in in vitro assay.

Mifepristone (RU486) is a progesterone receptor (PR) and glucocorticoid receptor (GR) antagonist with IC50s of 0.2 nM and 2.6 nM in in vitro assay.

Specifications

Chemical Name or Material Mifepristone
Physical Form Powder
CAS 84371-65-3
Color Light Yellow
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C29H35NO2
For Use With (Application) Cancer-programmed cell death
Synonym RU486 RU 38486
Solubility Information DMSO : 100 mg/mL (232.78 mM; ultrasonic and warming and heat to 60°C)
SMILES C[C@@]12[C@@](C#CC)(O)CC[C@@]1([H])[C@]3([H])CCC4=CC(CCC4=C3[C@@H](C5=CC=C(N(C)C)C=C5)C2)=O
Molecular Weight (g/mol) 429.59
Formula Weight 429.59
Percent Purity 99.77%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.