missing translation for 'onlineSavingsMsg'
Learn More

MIF4GD Antibody (OTI4C11), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18418216 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418216 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418216 Supplier Novus Biologicals Supplier No. NBP272685MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MIF4GD Monoclonal antibody specifically detects MIF4GD in Human, Mouse, Rat samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIF4GD
Applications Flow Cytometry
Classification Monoclonal
Clone OTI4C11
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias AD023, hSLIP1, MGC45027, MIF4G domain containing, MIFD, SLBP (stem loop binding protein)-interacting protein 1, SLBP-interacting protein 1, SLIP1MIF4G domain-containing protein
Host Species Mouse
Immunogen Full length human recombinant protein of human MIF4GD(NP_065730) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57409
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.