missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIER1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11452-1-AP-20UL
This item is not returnable.
View return policy
Description
The association of hMI-ER1 with Sp1 represents a novel mechanism for the negative regulation of Sp1 target promoters. Results demonstrate that alternate use of a facultative intron regulates the subcellular localization of hMI-ER1 proteins and this may have important implications for hMI-ER1 function in the regulation of transcription. Repressor activity is due to interaction and recruitment of a trichostatin A-sensitive histone deacetylase 1 (HDAC1).Specifications
| MIER1 | |
| Polyclonal | |
| Unconjugated | |
| MIER1 | |
| Early response 1, ER1, hMI ER1, KIAA1610, MI ER1, MIER1, RP5 944N15.1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 313418, 57708, 71148 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.44 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5UAK0, Q8N108 | |
| Mier1 | |
| MIER1 Fusion Protein Ag2010 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction