Learn More
MIER1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554032
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MMEGETNFSS EIEDLAREGD MPIHELLSLY GYGSTVRLPE EDEEEEEEEE EGEDDEDADN DDNSGCSGEN KEENIKDSSG QEDETQSSND DPSQSVASQD AQEI Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.
The association of hMI-ER1 with Sp1 represents a novel mechanism for the negative regulation of Sp1 target promoters. Results demonstrate that alternate use of a facultative intron regulates the subcellular localization of hMI-ER1 proteins and this may have important implications for hMI-ER1 function in the regulation of transcription. Repressor activity is due to interaction and recruitment of a trichostatin A-sensitive histone deacetylase 1 (HDAC1).Specifications
MIER1 | |
Polyclonal | |
Unconjugated | |
MIER1 | |
4933425I22Rik; 5830411K19Rik; early response 1; er1; hMI-ER1; Kiaa1610; mesoderm induction early response 1 homolog; mesoderm induction early response 1 homolog (Xenopus laevis; mesoderm induction early response 1, transcriptional regulator; mesoderm induction early response protein 1; MIER1; MI-ER1; MIER1 transcriptional regulator; RGD1562337; RP5-944N15.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57708 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N108 | |
MIER1 | |
Recombinant protein corresponding to Human MIER1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only