Learn More
Miele Professional™ Miele Process Documentation
Crossover network cable for direct connection to PC, length 5 m (type 9).
Brand:  Miele Professional™ 7951500
Additional Details : Weight : 0.15000kg
Product Code. 15490455
  • £46.35 / Each
Estimated Shipment: 12-08-2024
to see stock.
Add to basket
Description
Description
  • Cable category 6
  • Color: Transparent silver
  • Used for the network connection of Melag sterilizers
Specifications
Specifications
Process/Operating Documentation/Communication
Washer-disinfectors, laboratory