missing translation for 'onlineSavingsMsg'
Learn More

Midodrine hydrochloride, MedChemExpress

Product Code. 16491517
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16491517

Brand: Medchem Express HY12749A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Midodrine hydrochloride ((±)-Midodrine hydrochloride) is an α1-receptor agonist, for the treatment of dysautonomia and orthostatic hypotension.

Midodrine hydrochloride ((±)-Midodrine hydrochloride) is an α1-receptor agonist, for the treatment of dysautonomia and orthostatic hypotension.

Specifications

Chemical Name or Material Midodrine hydrochloride
CAS 43218-56-0
Color Light Brown
Purity Grade Notes Research
Molecular Formula C12H19ClN2O4
Quantity 50 mg
Synonym (±)-Midodrine hydrochloride
Solubility Information H2O : 100 mg/mL (343.95 mM; Need ultrasonic)
SMILES O=C(NCC(C1=CC(OC)=CC=C1OC)O)CN.[H]Cl
Molecular Weight (g/mol) 290.74
Formula Weight 290.74
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.