missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MIDN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561968
This item is not returnable.
View return policy
Description
Immunogen sequence: LNDLLSATRH YQGMPPSLAQ LRCHAQCSPA SPAPDLAPRT TSCEKLTAAP SASLLQGQSQ IRMCKPPGDR LRQTENRATR CKVER Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 88%.
Expression of the midnolin gene is developmentally regulated: it is strongly expressed at the mesencephalon (midbrain) of the embryo in day 12.5 (E12.5) mice. The midnolin encodes a protein of 508 amino acids (aa), which contains a Ubiquitin-like domain. The intracellular distribution of the midnolin was studied by using midnolin-green fluorescent protein (GFP) fusion proteins. Midnolin was found to be localized in the nucleus and nucleolus, but not in the cytoplasm. The nucleolar localization signal was determined to be a 28aa peptide located at the C-terminal region of the midnolin. May be involved in regulation of genes related to neurogenesis in the nucleolus.
Specifications
| MIDN | |
| Polyclonal | |
| Unconjugated | |
| MIDN | |
| 3000003C15Rik; DKFZp547M072; midbrain nucleolar protein; Midn; Midnolin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90007 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q504T8 | |
| MIDN | |
| Recombinant protein corresponding to Human MIDN. Recombinant protein control fragment (Product #RP-99142). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction