Learn More
PeproTech Midkine Polyclonal Antibody, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
£177.00 - £1468.00
Specifications
| Antigen | Midkine |
|---|---|
| Concentration | 0.1-1.0 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
17814613
![]() |
PeproTech
500-P171-50UG |
50 μg |
£177.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17883833
![]() |
PeproTech
500-P171-100UG |
100 μg |
£228.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17844413
![]() |
PeproTech
500-P171-1MG |
1 mg |
£1468.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
AA Sequence of recombinant protein: VAKKKDKVKK GGPGSECAEW AWGPCTPSSK DCGVGFREGT CGAQTQRIRC RVPCNWKKEF GADCKYKFEN WGACDGGTGT KVRQGTLKKA RYNAQCQETI RVTKPCTPKT KAKAKAKKGK GKD. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Midkine. Anti-Human Midkine-specific antibody was purified by affinity chromatography employing an immobilized Human Midkine matrix. Sandwich ELISA: To detect Human Midkine by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human Midkine (500-P171Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Midkine. Western Blot: To detect Human Midkine by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human Midkine is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Midkine is a retinoic acid-responsive, heparin-binding growth factor expressed in various cell types during embryogenesis. It promotes angiogenesis, cell growth, and cell migration. Midkine is also expressed in several carcinomas, suggesting that it may play a role in tumorigenesis, perhaps through its effects on angiogenesis.Specifikationer
| Midkine | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P21741 | |
| 4192 | |
| E.coli-derived Recombinant Human Midkine. | |
| Antigen affinity chromatography | |
| Mdk |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| amphiregulin-associated protein; ARAP; FLJ27379; MDK; Mek; Midgestation and kidney protein; midkine; midkine (neurite growth-promoting factor 2); MK; MK1; NEGF2; Neurite outgrowth-promoting factor 2; Neurite outgrowth-promoting protein; Retanoic acid-responsive protein; retinoic acid inducible factor; retinoic acid-induced differentiation factor; retinoic acid-responsive protein | |
| Mdk | |
| Primary | |
| -20°C |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.