Learn More
Invitrogen™ Midkine Monoclonal Antibody (1011522)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA561180
Product Code. 30533856
  • £357.00 / 100µg
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Midkine is a retinoic acid-responsive, heparin-binding growth factor expressed in various cell types during embryogenesis. It promotes angiogenesis, cell growth, and cell migration. Midkine is also expressed in several carcinomas, suggesting that it may play a role in tumorigenesis, perhaps through its effects on angiogenesis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Midkine
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P21741
Mdk
E. coli-derived recombinant human Midkine, Lys23-Asp143.
100 μg
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry
1011522
Unconjugated
Mdk
amphiregulin-associated protein; ARAP; FLJ27379; MDK; Mek; Midgestation and kidney protein; midkine; midkine (neurite growth-promoting factor 2); MK; MK1; NEGF2; Neurite outgrowth-promoting factor 2; Neurite outgrowth-promoting protein; Retanoic acid-responsive protein; retinoic acid inducible factor; retinoic acid-induced differentiation factor; retinoic acid-responsive protein
Mouse
Protein A/G
RUO
4192
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized