missing translation for 'onlineSavingsMsg'
Learn More

Christeyns™ Mida Chriox 5

Product Code. 17831445
Change view
Click to view available options
Format:
Can
Drum
Size:
22 kg
222 kg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Size unitSize
17831445 Can 22 kg Each
17841445 Drum 222 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 17831445 Supplier Christeyns™ Supplier No. 695935
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Mida Chriox 5 is a biodegradable, low residue broad spectrum sporicidal made of peracetic acid.

  • Disinfectant based on peracetic acid
  • Recommended for hygiene in cosmetic and pharmaceutical industries
  • Suitable for the organic and cosmetic industries
  • Usable in CIP without surfactants
  • Measurement and control by conductivity
  • Authorized for the following applications in: TP2: Disinfectant used in the private sector and in the field of public health and other biocidal products; TP4: Disinfectant for surfaces in contact with food and animal feed with mandatory rinsing

Specifications

Product Type Disinfectant Concentrate
Color-coded No
Form Concentrated Liquid
pH Neutral
Scent Regular scent
Sterility Non-sterile
Format Can
Size 22 kg
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mida CHRIOX 5
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contain: hydrogen peroxide, acetic acid, peracetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.