missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MID2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £375.00
Specifications
| Antigen | MID2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18323377
|
Novus Biologicals
NBP3-05060-20UL |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18326176
|
Novus Biologicals
NBP3-05060-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MID2 Polyclonal specifically detects MID2 in Human, Mouse samples. It is validated for Western Blot.Spécification
| MID2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| EC 6.3.2.-, FLJ37715, FLJ41813, FXY2midin-2, midin 2, Midin-2, midline 2, Midline defect 2, Midline-2, RING finger protein 60, RNF60midline-2, TRIM1probable E3 ubiquitin-protein ligase MID2, tripartite motif protein 1, Tripartite motif-containing protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 476-705 of human MID2 (NP_438112.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Zinc Finger | |
| PBS with 50% glycerol, pH7.3. | |
| 11043 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title