missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MID1IP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15764-1-AP-150UL
This item is not returnable.
View return policy
Description
Plays a role in the regulation of lipogenesis in liver. Up-regulates ACACA enzyme activity. Required for efficient lipid biosynthesis, including triacylglycerol, diacylglycerol and phospholipid. Involved in stabilization of microtubules.Specifications
| MID1IP1 | |
| Polyclonal | |
| Unconjugated | |
| MID1IP1 | |
| G12 like, Mid1 interacting protein 1, MID1IP1, MIG12, Protein STRAIT11499, STRAIT11499, THRSPL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 404280, 58526, 68041 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6P7D5, Q9CQ20, Q9NPA3 | |
| Mid1ip1 | |
| MID1IP1 Fusion Protein Ag8400 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction