missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MID1IP1 Rabbit anti-Human, Clone: 5, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90264-100UL
This item is not returnable.
View return policy
Description
MID1IP1 Monoclonal specifically detects MID1IP1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| MID1IP1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| 5 | |
| Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunofluorescence | |
| FLJ10386, G12-like, Gastrulation-specific G12-like protein, MID1 interacting protein 1 (gastrulation specific G12 homolog (zebrafish)), MID1 interacting protein 1 (gastrulation specific G12-like (zebrafish)), MID1 interacting protein 1 (gastrulation specific G12-like), Mid1-interacting G12-like protein, mid1-interacting protein 1, MIG12MID1 interacting G12-like protein, Protein STRAIT11499, S14R, Spot 14-R, Spot 14-related protein, STRAIT11499, THRSPL | |
| Recombinant Human MID1IP1 Protein | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 58526 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering